missing translation for 'onlineSavingsMsg'
Learn More

B7-H7/HHLA2 Antibody (907812R) [Janelia Fluor™ 525], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB80841RJF525

4364.75 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 30170945

  • 5075.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

B7-H7/HHLA2 Monoclonal antibody specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

B7-H7/HHLA2
Monoclonal
Janelia Fluor 525
HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44
RUO
Primary
Human
Liquid
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
907812R
50mM Sodium Borate
Mouse
0.1 mL
Immunology, Inflammation
11148
Store at 4°C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.