missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (16-10A1) [CoraFluor™ 1], Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-43385CL1
7463.00 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Feline,Human,Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Frozen)
Specifications
| B7-1/CD80 | |
| Monoclonal | |
| CoraFluor 1 | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| The immunogen for this antibody was CD80. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| 16-10A1 | |
| PBS | |
| Armenian Hamster | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Feline, Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction