missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568109
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse samples.
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene encodes a protein that functions in N-linked glycoprotein glycosylation and shows strict donor substrate specificity for UDP-galactose. [provided by RefSeq, Jul 2008].Specifications
B3GALT2 | |
Polyclonal | |
Unconjugated | |
B3GALT2 | |
BETA3GALT2; GLCT2; beta3Gal-T2; beta-1,3-galactosyltransferase 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2; UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2; beta-1,3-GalTase 2; beta-3-galt2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26878, 8707 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43825, O54905 | |
B3GALT2 | |
Synthesized peptide derived from human B3GALT2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |