Learn More
Thermo Scientific Chemicals Aztreonam
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Type | Aztreonam |
Melting Point | 227.0°C |
Ignition Residue | 0.1% max. |
Infrared Spectrum | Authentic |
Solubility Information | Solubility in water: very slightly soluble. Other solubilities: very slightly soluble in methanol, soluble in dmso, insoluble in ethanol |
Formula Weight | 435.43 |
Percent Purity | ≥90% (HPLC) |
Water | 5% max. |
Quantity | 1 g |
Chemical Name or Material | Aztreonam |