missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1F Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17725-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847674

  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.
Specifications

Specifications

ATP6V1F
Polyclonal
Unconjugated
ATP6V1F
ATP6S14, ATP6V1F, V ATPase 14 kDa subunit, V ATPase subunit F, V type proton ATPase subunit F, Vacuolar proton pump subunit F, VATF, Vma7
Rabbit
Antigen Affinity Chromatography
RUO
66144, 9296
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16864, Q9D1K2
Atp6v1f
ATP6V1F Fusion Protein Ag12121
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers