Learn More
Trajan Scientific and Medical™ ATLAS Optic Frit and Tapered Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
615.00 NOK - 2425.00 NOK
Specifications
Length (Metric) | 81 mm |
---|---|
Diameter (Metric) Inner | 3 mm |
Diameter (Metric) Outer | 5 mm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12934091
|
Trajan Scientific and Medical™
092273 |
5/Pk. |
2425.00 NOK
Pack of 5 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
12944091
|
Trajan Scientific and Medical™
09227301 |
1 Ea. |
615.00 NOK
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with ATAS Optic instruments
Specifications
81 mm | |
3 mm | |
5 mm |