missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Asunaprevir
GREENER_CHOICE

Product Code. 18758066
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18758066

Brand: Tocris Bioscience™ 7755/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent hepatitis C virus (HCV) NS3 protease inihibitor

Asunaprevir is a potent HCV NS3 protease inhibitor (IC50 = 0.2-3.5 nM). Asunaprevir can be used to control CRIPSR-Cas9 gene editing via the small molecule-assisted shut-off (SMASh) technique, which involves fusion of Cas9 to a peptide comprising a protease domain and a degron tag. Asuprenavir blocks the protease-mediated removal of the degron tag leading to degradation of Cas9 through proteasome and autophagolysosome pathways. It can also be used as a resversible switch to control CAR (chimeric antigen receptor)-T cells, where the CAR has been modified by insertion of a hepatitis C virus nonstructural protein 3 protease (HCV-NS3) between the scFv and the hinge domains. Also inhibits SARS-CoV-2 Mpro activity (IC50 = 15-48.4 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[[N-tert-Butoxycarbonyl-L-tert-leucyl-trans-4-[(7-chloro-4-methoxyisoquinolin-1-yl)oxy]-L-prolyl]amino]-N-(cyclopropylsulfonyl)-(1 R,2 S)-2-vinylcyclopropane-1-carboxamide
CAS 630420-16-5
Quantity 10 mg
Target CRISPR Reagents
Molecular Formula C35H46ClN5O9S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.