missing translation for 'onlineSavingsMsg'
Learn More

Asunaprevir, MedChemExpress

Product Code. 16418677
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418677

Brand: Medchem Express HY14434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asunaprevir (BMS-650032) is a potent and orally bioavailable hepatitis C virus (HCV) NS3 protease inhibitor, with IC50 of 0.2 nM-3.5 nM. Asunaprevir inhibits SARS-CoV-2 3CLpro activity.

Asunaprevir (BMS-650032) is a potent and orally bioavailable hepatitis C virus (HCV) NS3 protease inhibitor, with IC50 of 0.2 nM-3.5 nM. Asunaprevir inhibits SARS-CoV-2 3CLpro activity.

Specifications

Chemical Name or Material Asunaprevir
CAS 630420-16-5
Color White
Purity Grade Notes Research
Molecular Formula C35H46ClN5O9S
Quantity 5 mg
Synonym BMS-650032
Solubility Information DMSO : ≥ 100 mg/mL (133.64 mM) ∣Ethanol : 20 mg/mL (26.73 mM; Need ultrasonic)
SMILES ClC1=CC=C2C(C(O[C@H]3CN(C([C@@H](NC(OC(C)(C)C)=O)C(C)(C)C)=O)[C@H](C(N[C@@]4(C(NS(C5CC5)(=O)=O)=O)[C@H](C=C)C4)=O)C3)=NC=C2OC)=C1
Molecular Weight (g/mol) 748.29
Formula Weight 748.29
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.