missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Astrovirus type 2 Antibody (8-E7) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-63066JF669
This item is not returnable.
View return policy
Description
Astrovirus type 2 Monoclonal antibody specifically detects Astrovirus type 2 in Virus samples. It is validated for ELISA,Immunoprecipitation
Specifications
| Astrovirus type 2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Immunoprecipitation | |
| 8-E7 | |
| 50mM Sodium Borate | |
| Astrovirus type 2 expressed in LLC MK2 cells. | |
| 0.5 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction