missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11934-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ADP-ribosylation factors (ARFs) are highly conserved guanine nucleotide binding proteins that enhance the ADP-ribosyltransferase activity of cholera toxin. ARFs are important in eukaryotic vesicular trafficking pathways and they play an essential role in the activation of phospholipase D (PC-PLD). ARL15 (ADP-ribosylation factor-like 15), is a member of the ARF family of the Ras superfamily of small GTPases that are known to be involved in multiple regulatory pathways altered in human carcinogenesis. Other members of this protein family include ARL11, thought to be a tumor suppressor and may play a role in the regulation of apoptosis, ARL2 which plays an important role in microtubule dynamics and cell cycle progression, and ARL4, which may play a role in neurogenesis during embryonic development. The gene encoding ARL15 maps to chromosome 5p15 in humans. At least three isoforms of ARL15 are known to exist.Specifications
ARL15 | |
Polyclonal | |
Unconjugated | |
ARL15 | |
ARF related protein 2, ARFRP2, ARL15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218639, 54622, 689079 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BGR6, Q9NXU5 | |
ARL15 | |
ARL15 Fusion Protein Ag2530 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |