Learn More
Arginase 1 Monoclonal Antibody (A1exF5), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1434.00 NOK - 3300.00 NOK
Specifications
Antigen | Arginase 1 |
---|---|
Clone | A1exF5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933244
|
Affymetrix eBioscience
25-3697-80 |
25 μg |
1434.00 NOK
25µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943244
|
Affymetrix eBioscience
25-3697-82 |
100 μg |
3300.00 NOK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody A1exF5 recognizes both human and mouse Arginase 1, a cytosolic enzyme (Arg1). This A1exF5 clone is compatible with both, the standard intracellular protocols, and the Foxp3/Transcription Factor Staining Buffer Set. Applications Reported: This A1exF5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A1exF5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product No. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.
Arginase 1 Monoclonal antibody specifically detects Arginase 1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Arginase 1 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase | |
ARG1 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
A1exF5 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Human, Mouse | |
P05089, Q61176 | |
11846, 383 | |
E.coli-derived recombinant mouse Arginase 1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ARG1 |