missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ARF3 (Human) Recombinant Protein
Human ARF3 full-length ORF ( AAH07762, 1 a.a. - 181 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000377-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGNIFGNLLKSLIGKKEMRILMVGLDAAGKTTILYKLKLGEIVTTIPTIGFNVETVEYKNISFTVWDVGGQDKIRPLWRHYFQNTQGLIFVVDSNDRERVNEAREELMRMLAEDELRDAVLLVFANKQDLPNAMNAAEITDKLGLHSLRHRNWYIQATCATSGDGLYEGLDWLANQLKNKK
Specifications
AAH07762 | |
ADP-ribosylation factor 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
377 | |
Wheat germ expression system | |
25 μg | |
ARF3 | |
GST |