Learn More
Uvex™ Arc Protect Safety Glove
Click to view available options
Size:
7
8
9
10
11
The Uvex arc protect g1 protective glove provides safe protection against the thermal discharge of a short circuit electric arc.
  • The glove's high level of flexibility offers pleasant wearer comfort and an excellent tactile feel
  • Thermal protection
  • Certified in line with OEKO-TEX Standard 100
  • Protective glove made of modacrylic/cotton knit
  • Ergonomic fit
  • Excellent wearer comfort
  • Outstanding tactile feel
  • Excellent protection against thermal discharge
Certifications/Compliance EN 61482-1-2, EN 407
Color Gray
Material Cotton, Modacryl
Size 8