Learn More
Duran™ AR Glass Disposable Culture Tubes
Disposable culture tubes for general purpose. DURAN™ AR Glass Disposable Culture Tubes are used in the laboratory setting.
95.80 NOK - 585.00 NOK
Specifications
Material AR Glass
Product Type Disposable Culture Tube
View More Specs
Products 16
Product Code Brand Capacity (Metric) Length (Metric) Diameter (Metric) Outside Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Diameter (Metric) Outside Price Quantity & Availability  
10595272 View Documents Duran™
231720784
8mL 100mm 11.75mm 560.00 NOK
Pack of 550 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10766771 View Documents Duran™
231721197
9mL 100mm 12.25mm 315.00 NOK
Pack of 500 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10223311 View Documents Duran™
231751159
6mL 100mm 382.00 NOK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10431541 View Documents Duran™
231751459
12mL 100mm 438.00 NOK
Pack of 100 View alternative products
Save Up to 
»
In Stock 57
Add to basket
15213035 View Documents Duran™
231721283
18mL 120mm 12.25mm 251.00 NOK
Pack of 250 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
15299574 View Documents Duran™
231721489
14mL 100mm 15.5mm 197.00 NOK
Pack of 250 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10739211 View Documents Duran™
231752155
22mL 505.00 NOK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
16272261 View Documents Duran™
231721986
19mL 150mm 15.5mm 259.00 NOK
Pack of 250 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10643982 View Documents Duran™
231752352
32mL 180mm 18mm 585.00 NOK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
15267556 View Documents Duran™
231720365
4mL 75mm 10mm 95.80 NOK
Pack of 250 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10208801 View Documents Duran™
231720587
5mL 75mm 11.75mm 268.00 NOK
Pack of 550 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10598411 View Documents Duran™
231720965
7mL 75mm 12.25mm, wall thickness: 0.55mm 202.00 NOK
Pack of 500 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10716961 View Documents Duran™
231720998
7mL 75mm 12.25mm, wall thickness: 0.8mm 283.00 NOK
Pack of 500 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
10320542 View Documents Duran™
231720184
4mL 75mm 9.75mm 365.00 NOK
Pack of 814 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
15666925 View Documents Duran™
231721986
19mL 150mm 15.5mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +47 22 95 59 51 for assistance
N/A View alternative products
Save Up to 
»
10060371 View Documents Duran™
231721094
15mL 100mm 15.75mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +47 22 95 59 51 for assistance
N/A View alternative products
Save Up to 
»
Specifications
Specifications
AR Glass
Disposable Culture Tube