missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Aqua Comparitor Color Discs: Phosphate
For testing water with the Aqua Comparator
301.00 NOK
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
11705458
|
Lovibond™
146250 |
Phosphate HR VANA Color Disc | 0 to 80mg/L |
301.00 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
11795448
|
Lovibond™
146240 |
Phosphate Color Disc | 0 to 4mg/L |
301.00 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for phosphates using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ |