missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13177-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene interacts with the cytoplasmic domains of amyloid beta precursor protein and amyloid beta precursor-like protein 2. This protein contains two phosphotyrosine binding domains, which are thought to function in signal transduction. Polymorphisms in this gene have been associated with Alzheimer′s disease. Alternative splicing results in multiple transcript variants.Specifications
APBB2 | |
Polyclonal | |
Unconjugated | |
APBB2 | |
APBB2, FE65L, FE65L1, Protein Fe65 like 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11787, 305338, 323 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92870, Q9DBR4 | |
APBB2 | |
APBB2 Fusion Protein Ag3844 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |