missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STUB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535110
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken and mouse based on sequence homology.
STUB1 Polyclonal antibody specifically detects STUB1 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
STUB1 | |
Polyclonal | |
Unconjugated | |
STUB1 | |
0610033N24Rik; 2210017D18Rik; 2310040B03Rik; Antigen NY-CO-7; AW046544; Carboxy terminus of Hsp70-interacting protein; CHIP; CLL-associated antigen KW-8; E3 ubiquitin-protein ligase CHIP; heat shock protein A binding protein 2 (c-terminal); HSPABP2; LA16c-313D11.6; NY-CO-7; PP1131; RING-type E3 ubiquitin transferase CHIP; SCAR16; SDCCAG7; serologically defined colon cancer antigen 7; STIP1 homology and U box-containing protein 1; STIP1 homology and U-box containing protein 1; STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase; Stub1; UBOX1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10273 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9UNE7 | |
STUB1 | |
KLH conjugated synthetic peptide between 236-263 amino acids from the C-terminal region of human STUB1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |