Learn More
SSTR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3111
Additional Details : Weight : 1.24740kg
Description
PA3-111 detects the Somatostatin Receptor 4 in human and mouse samples. PA3-111 has been used successfully in Western blot, immunocytochemistry, and immunohistochemistry procedures. In Western blot analysis of SSTR4 transfected HEK-293 cells this antibody detects a ∽40-50 kDa protein representing SSTR4. The PA3-111 immunogen is a synthetic peptide corresponding to residues C(366) Q Q E A L Q P E P G R K R I P L T R T T T F(388) of human SSTR4.
SSTR4 Polyclonal antibody specifically detects SSTR4 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SSTR4 | |
Polyclonal | |
Unconjugated | |
SSTR4 | |
G-protein coupled receptor; Smstr4; somatostatin receptor; somatostatin receptor 4; Somatostatin receptor subtype 4 Rattus norvegicus Sprague-Dawley major hippocampal somatostatin receptor (SSTR4) mRNA; somatostatin receptor type 4; SS4R; SS-4-R; SS4-R; sst4; Sstr4 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P31391, P49660 | |
SSTR4 | |
Synthetic peptide corresponding to residues C(366) Q Q E A L Q P E P G R K R I P L T R T T T F(388) of human SSTR4. | |
RUO | |
20608, 6754 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |