Learn More
PCNA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527214
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, PC-12, A549, SiHa. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (92%), Xenopus laevis (89%), Pig (100%), Chicken (96%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PCNA Polyclonal antibody specifically detects PCNA in Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot.Specifications
PCNA | |
Polyclonal | |
Unconjugated | |
Pcna | |
ATLD2; cb16; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; fa28e03; fb36g03; HGCN8729; hypothetical protein LOC515499; MGC8367; Pcna; pcna protein; Pcna/cyclin; PCNAR; POL30; Proliferating cell nuclear antigen; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100689335, 18538, 25737, 5111 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot | |
0.11 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P04961, P12004, P17918, P57761 | |
Pcna | |
Recombinant fragment corresponding to a region within amino acids 1 and 253 of Human PCNA | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Rat | |
Antibody | |
IgG |