Learn More
p27 Kip1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527188
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, MCF-7, MDA-MB-231, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12. Predicted reactivity: Mouse (80%), Dog (87%), Cat (87%), Pig (84%), Rhesus Monkey (98%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
p27 Kip1 Polyclonal antibody specifically detects p27 Kip1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
p27 Kip1 | |
Polyclonal | |
Unconjugated | |
Cdkn1b | |
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1027, 12576 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.08 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P46414, P46527 | |
Cdkn1b | |
Recombinant fragment corresponding to a region within amino acids 55 and 198 of Human P27 Kip1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |