missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxytocin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526701
Additional Details : Weight : 0.01000kg
Description
Oxytocin Polyclonal antibody specifically detects Oxytocin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Oxytocin | |
Polyclonal | |
Unconjugated | |
OXT | |
neurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein product | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P01178 | |
OXT | |
KLH conjugated synthetic peptide between 11-40 amino acids from the central region of human NEU1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |