Learn More
Nectin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529757
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, MCF-7. Predicted reactivity: Rhesus Monkey (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Nectin 2 Polyclonal antibody specifically detects Nectin 2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Nectin 2 | |
Polyclonal | |
Unconjugated | |
NECTIN2 | |
AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5819 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q92692 | |
NECTIN2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 239 of Human Nectin 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |