Learn More
Mouse IgM Isotype Control, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen PA533228
Additional Details : Weight : 0.01000kg
Description
PA5-33228 may be used as an isotype matched negative control to determine the level of non-specific binding of IgM monoclonal antibodies in human leucocytes. The antibody does not recognize human antigen. This antibody recognizes a rat cell surface marker and therefore cannot be used as a negative control in the species. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgM Control antibody specifically detects Mouse IgM in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgM | |
0.1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
Mouse IgM | |
100 Tests | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgM |