missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-108 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543523
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: NFITWLFNET SLAFIVPHET KSPEIHVTNP KQGKRLNFTQ SYSLQLSNLK Sequence homology: Human: 100%
Ly-108 Polyclonal antibody specifically detects Ly-108 in Human samples. It is validated for Western Blot.Specifications
Ly-108 | |
Polyclonal | |
Unconjugated | |
SLAMF6 | |
Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
114836 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96DU3 | |
SLAMF6 | |
Synthetic peptide directed towards the N-terminal of human SLAMF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |