missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547927
Additional Details : Weight : 0.00010kg
Description
In Western blots, less than 1% cross-reactivity with recombinant human IL-7 R alpha is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD127 Polyclonal antibody specifically detects CD127 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD127 | |
Polyclonal | |
Unconjugated | |
Il7r | |
CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R a; IL 7Ra; IL 7Ra; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R a; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Ra; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16197 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P16872 | |
Il7r | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-7 R alpha/CD127 Glu21-Asp239 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |