Learn More
CD127 Monoclonal Antibody (40131), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523825
2450.12 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00010kg
Description
In Western blots, approximately 5% cross-reactivity was observed with recombinant human (rh) IL-10 R, rhIL-2 R beta, rhIL-5 R alpha, and rhIL-6 R. Reconstitute at 0.5 mg/mL in sterile PBS.
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD127 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P16871 | |
Il7r | |
Mouse myeloma cell line NS0-derived recombinant human IL-7 R alpha Extracellular domain | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
40131 | |
Unconjugated | |
Il7r | |
CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R a; IL 7Ra; IL 7Ra; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R a; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Ra; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
Mouse | |
Protein A/G | |
RUO | |
3575 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |