missing translation for 'onlineSavingsMsg'
Learn More

IL5RA Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen PA547340

Additional Details : Weight : 0.00010kg

Product Code. 15547832

  • 4125.00 NOK / 100µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) IL-1 RII, rhIL-2 R beta, rhIL-2 R gamma, rhIL-3 R is observed and less than 1% cross-reactivity with rhIL-1 RI, rhIL-4 R, rhIL-6 R, rhIL-7 R, rhIL-9 R, and rhIL-10 R is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IL5RA Polyclonal antibody specifically detects IL5RA in Human samples. It is validated for Flow Cytometry, Neutralization, Western Blot.
Specifications

Specifications

IL5RA
Polyclonal
Unconjugated
IL5RA
CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha
Goat
Antigen affinity chromatography
RUO
3568
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Flow Cytometry, Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q01344
IL5RA
Sf 21-derived recombinant human IL-5R alpha/CD125
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers