Learn More
IL-22 Receptor alpha Monoclonal Antibody (305405), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524017
2336.78 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10, rhIL-22BP or rhIL-20 R alpha is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
IL-22 Receptor alpha Monoclonal antibody specifically detects IL-22 Receptor alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
IL-22 Receptor alpha | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8N6P7 | |
Il22ra1 | |
CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
305405 | |
Unconjugated | |
Il22ra1 | |
9130219A07Rik; CRF2-9; Cytokine receptor class-II member 9; cytokine receptor family 2 member 9; ENSMUSG00000073746; il-22 receptor; IL-22 receptor subunit alpha-1; IL22R; IL-22R; IL22R1; IL22RA1; IL-22RA1; IL-22R-alpha-1; ILN; Interleukin; interleukin 22 receptor subunit alpha 1; interleukin 22 receptor, alpha 1; interleukin-22 receptor alpha chain; interleukin-22 receptor subunit alpha-1; RGD1559655; zcytoR11 | |
Mouse | |
Protein A/G | |
RUO | |
58985 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |