Learn More
Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21433
1321.39 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00570kg
Description
Product will be shipped at Room Temperature.
Human IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (H+L) Cross-Adsorbed in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Human IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 555 | |
Goat | |
purified | |
RUO | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |