missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1104
Additional Details : Weight : 0.10000kg
Description
GATA6 Polyclonal antibody specifically detects GATA6 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GATA6 | |
Polyclonal | |
Unconjugated | |
GATA6 | |
AA410133; ASD9; AVSD5; DNA-binding protein GATA-GT2; GATA binding protein 6; Gata6; GATA-6; GATA-binding factor 6; GATA-binding protein 6; LOW QUALITY PROTEIN: transcription factor GATA-6; transcription factor GATA6; transcription factor GATA-6; zinc-finger transcription factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14465, 2627 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q61169, Q92908 | |
GATA6 | |
Synthetic peptide sequence KYSGQDGLYIGVSLASPAE & KRFGAAGADASDSRAFPARE | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- GATA6 Polyclonal Antibody
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : glycerol, sodium chloride