Learn More
GAS6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528227
Additional Details : Weight : 0.01000kg
Description
PA5-28227 targets GAS6 in IF and WB applications and shows reactivity with Human samples. The PA5-28227 immunogen is recombit fragment corresponding to a region within amino acids 283 and 621 of Human GAS6. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
GAS6 | |
Polyclonal | |
Unconjugated | |
Q14393 | |
GAS6 | |
Recombit fragment corresponding to a region within amino acids 283 and 621 of Human GAS6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1.02 mg/mL | |
GAS6 | |
AXLLG, AXSF, AXL receptor tyrosine kinase ligand, AXL stimulatory factor, growth arrest-specific protein 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2621 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |