missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544158
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AEKVLVTGGA GYIGSHTVLE LLEAGYLPVV IDNFHNAFRG GGSLPESLRR Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 91%; Pig: 91%; Rabbit: 92%; Rat: 91%
GALE Polyclonal antibody specifically detects GALE in Human samples. It is validated for Western Blot.Specifications
GALE | |
Polyclonal | |
Unconjugated | |
GALE | |
2310002A12Rik; AI323962; EGK_00373; galactose-4-epimerase, UDP; galactose-4-epimerase, UDP-; galactowaldenase; Gale; SDR1E1; short chain dehydrogenase/reductase family 1E, member 1; UDP galactose-4'-epimerase; UDP-galactosamine 4-epimerase; UDP-galactose 4-epimerase; UDP-galactose-4-epimerase; UDP-GalNAc 4-epimerase; UDP-GlcNAc 4-epimerase; UDP-glucose 4-epimerase; UDP-N-acetylgalactosamine 4-epimerase; UDP-N-acetylglucosamine 4-epimerase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2582 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14376 | |
GALE | |
Synthetic peptide directed towards the N-terminal of human GALE | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |