Learn More
Cytokeratin 19 Monoclonal Antibody (A53-B/A2.26 (Ks19.1)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512663
2751.02 NOK valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
MA5-12663 targets Cytokeratin 19 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The MA5-12663 immunogen is human breast cancer MCF-7 cells.
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Cytokeratin 19 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P08727 | |
KRT19 | |
Human breast cancer MCF-7 cells | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a λ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
A53-B/A2.26 (Ks19.1) | |
Unconjugated | |
KRT19 | |
40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 | |
Mouse | |
Protein G | |
RUO | |
3880 | |
4° C | |
Liquid |