missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRP Monoclonal Antibody (C7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110320
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes antigen both in the presence and in the absence (samples containing EDTA) of Ca2+.
CRP Monoclonal antibody specifically detects CRP in Human samples. It is validated for ELISA.Specifications
CRP | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P02741 | |
Crp | |
Purified human CRP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
C7 | |
Unconjugated | |
Crp | |
Aa1249; Ab1-341; Ab2-196; Ac1-114; Ac1262; Ac2-069; acute phase protein; CRP; AI255847; Ba2-693; c reactive; C-reactive preprotein; C-reactive protein; C-reactive protein member of the pentraxin family; C-reactive protein(1-205); C-reactive protein, member of the pentraxin family; C-reactive protein, pentraxin-related; C-reactive protein, petaxin related; CRP; CRP1; CSRP; CYRP; D1S181E; DKFZp686M148; DKFZp686M149; MGC149895; MGC88244; pentraxin 1; PTX1; RP11-419N10.4 | |
Mouse | |
Protein A | |
RUO | |
1401 | |
4° C, do not freeze | |
Liquid |