missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD98 Monoclonal Antibody (MEM-108), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119586
2023.43 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
CD98 Monoclonal antibody specifically detects CD98 in Human samples. It is validated for Flow Cytometry.Specifications
CD98 | |
Monoclonal | |
FITC | |
SLC3A2 | |
4F2; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2hc; 4T2HC; AI314110; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; Cd98; CD98 antigen; CD98 heavy chain; CD98HC; Ly10; Ly-10; Ly-m10; Lymphocyte activation antigen 4F2 large subunit; MDU1; Mgp-2hc; monoclonal antibody 44D7; NACAE; RGD:3073}; SLC3A2; slc3a2 {ECO:0000312; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; solute carrier family 3 member 2; solute carrier family 3, member 2; type II transmembrane protein | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
6520 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-108 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08195 | |
SLC3A2 | |
Burkitt's lymphoma cell line Raji. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |