missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Monoclonal Antibody (M38), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110292
2030.10 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
CD81 Monoclonal antibody specifically detects CD81 in Feline, Human, Rabbit samples. It is validated for Flow Cytometry.Specifications
CD81 | |
Monoclonal | |
PE | |
CD81 | |
26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
101093144, 975 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
M38 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P60033 | |
CD81 | |
MOLT-4 (human T-ALL cell line) | |
100 Tests | |
Primary | |
Feline, Human, Rabbit | |
Antibody | |
IgG1 |