missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD73 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511870
Additional Details : Weight : 0.01000kg
Description
CD73 Polyclonal antibody specifically detects CD73 in Human samples. It is validated for Immunohistochemistry.Specifications
CD73 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P21589 | |
NT5E | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CD73 (NT5E) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
NT5E | |
2210401F01Rik; 5 nucleotidase, ecto; 5' nucleotidase, ecto; 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; NT; Nt5; Nt5e; NTE; Purine 5-Prime-Nucleotidase | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
4907 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |