Learn More
CD72 Monoclonal Antibody (Bu40), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516599
Additional Details : Weight : 0.01000kg
Description
This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD72 Monoclonal antibody specifically detects CD72 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD72 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P21854 | |
CD72 | |
The details of the immunogen for this antibody are not currently available. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Bu40 | |
Unconjugated | |
CD72 | |
B-cell differentiation antigen CD72; CD72; CD72 antigen; Cd72 molecule; CD72b; CD72c; Ly-19; Ly32; Ly-32; LYB2; Lyb-2; Ly-m19; lymphocyte antigen 32; RP11-331F9.9 | |
Mouse | |
Protein A | |
RUO | |
971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |