missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD70 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532700
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Marmoset (94%) Bovine, Rabbit (82%).
CD70 Polyclonal antibody specifically detects CD70 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CD70 | |
Polyclonal | |
Unconjugated | |
Cd70 | |
CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P32970 | |
Cd70 | |
Synthetic 17 amino acid peptide from internal region of human CD70. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |