missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529574
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CEACAM3 Polyclonal antibody specifically detects CEACAM3 in Human samples. It is validated for Western Blot.Specifications
CEACAM3 | |
Polyclonal | |
Unconjugated | |
CEACAM3 | |
carcinoembryonic antigen CGM1; Carcinoembryonic antigen gene family (CGM1); carcinoembryonic antigen gene family 4; carcinoembryonic antigen gene family member 1; Carcinoembryonic antigen gene family member 4 / (carcinoembryonic antigene-related protein); carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen-related cell adhesion molecule 3; CD66d; CD66d antigen; CEA; cea12; CEACAM3; ceacam3 {ECO:0000312; Ceacam5; Cear; Cgm1; Cgm4; CGM4AA; EG384557; nonspecific cross-reacting antigen; pregnancy-specific glycoprotein 24; Psg24; RATCEAA; RATCGM4AA; RGD:2333}; W264; W282 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1084 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P40198 | |
CEACAM3 | |
Recombinant fragment corresponding to a region within amino acids 55 and 252 of Human CD66d | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |