Learn More
P-Selectin Monoclonal Antibody (Psel.KO.2.12), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181632
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD62P antibody, clone Psel.KO.2.12 recognizes the CD62P cell surface antigen, an approximately 140 kDa glycoprotein also known as P-selectin.
P-Selectin Monoclonal antibody specifically detects P-Selectin in Goat, Human, Ovine, Rat samples. It is validated for Flow Cytometry.Specifications
P-Selectin | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P16109, P98106, P98109 | |
SELP | |
CD62P transfected 300.19 cells. | |
100 Tests | |
Primary | |
Goat, Human, Ovine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
Psel.KO.2.12 | |
PE | |
SELP | |
CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp | |
Mouse | |
Protein A | |
RUO | |
102170076, 25651, 443305, 6403 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |