missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L Monoclonal Antibody (OX-85), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517572
Additional Details : Weight : 0.01000kg
Description
CD62L Monoclonal antibody specifically detects CD62L in Rat samples. It is validated for Flow Cytometry.Specifications
CD62L | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P30836 | |
SELL | |
Recombinant rat L-selectin | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-85 | |
Biotin | |
SELL | |
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
Mouse | |
Protein G | |
RUO | |
29259 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |