Learn More
CD4 Monoclonal Antibody (GK1.5), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA1146
1077.38 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Flow cytometry analysis of MA1-146 shows positive staining of mouse T-lymphocytes. The antibody also detects recombinant mouse CD4 at ∽55 kDa.
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
CD4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P06332 | |
CD4 | |
Mouse CTL clone V4 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
Flow Cytometry, Immunocytochemistry, Western Blot | |
GK1.5 | |
Unconjugated | |
CD4 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
Rat | |
Protein G | |
RUO | |
12504 | |
-20°C | |
Liquid |