Learn More
CD4/CD8 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA517009
3310.16 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
MA5-17009 contains a monoclonal mixture of FITC-conjugated CD4 (clone #YTS191.1, Rat IgG2b) and RPE-conjugated CD8 (clone #KT15, Rat IgG2a). This antibody is a dual-color reagent recognizing mouse CD4 and CD8 cell surface antigens. Clone YTS191.1 recognizes murine CD4, a T cell differentiation antigen expressed on thymocytes and helper/inducer T cells in the peripheral blood. Clone KT15 recognizes a non-polymorphic epitope on the murine CD8 alpha chain. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD4/CD8 Cocktail antibody specifically detects CD4/CD8 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD4/CD8 | |
Cocktail | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P01731, P06332 | |
CD4, CD8A | |
T cell clone, C6 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2a, IgG2b |
Flow Cytometry | |
YTS191.1/KT15 | |
FITC/PE | |
CD4 | |
BB154331; Cd4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; CD8; CD8 antigen; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8b; CD8b molecule; CD8B1; CD8beta; L3T4; LEU2; Leu-2; Leu2 T-lymphocyte antigen; Ly-2; LY3; Ly-35; Ly-4; Ly-B; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); LYT3; MAL; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; T-lymphocyte differentiation antigen T8/Leu-2 | |
Rat | |
purified | |
RUO | |
12504, 12525 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |