missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4/CD27 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA112121
Additional Details : Weight : 0.01000kg
Description
This antibody is a monoclonal mixture of FITC-conjugated CD4 (clone 7E14, isotype IgG1) and PE-conjugated CD27 (clone DEN-3, isotype IgG1).
CD4/CD27 Cocktail antibody specifically detects CD4/CD27 in Human samples. It is validated for Flow Cytometry.Specifications
CD4/CD27 | |
Cocktail | |
FITC/PE | |
CD4 | |
CD27; CD27 antigen; CD27 molecule; CD27L receptor; CD4; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; TNFRSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Mouse | |
Protein A/G | |
RUO | |
920, 939 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
7E14, DEN-3, DEN-3 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P01730, P26842 | |
CD27, CD4 | |
human peripheral blood T lymphocytes | |
50 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |