missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4/CD26 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA112118
Additional Details : Weight : 0.01000kg
Description
This antibody is a monoclonal mixture of FITC-conjugated CD4 (clone 7E14, isotype IgG1) and PE-conjugated CD26 (clone EUG-6, isotype IgG2a).
CD4/CD26 Cocktail antibody specifically detects CD4/CD26 in Human samples. It is validated for Flow Cytometry.Specifications
CD4/CD26 | |
Cocktail | |
FITC/PE | |
DPP4 | |
ADABP; ADCP2; ADCP-2; Adenosine deaminase complexing protein 2; CD26; CD4; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; dipeptidyl peptidase 4; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; DPPIV; DPP-IV; T-cell activation antigen CD26; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; TP103 | |
Mouse | |
Protein A/G | |
RUO | |
1803, 920 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
7E14, EUG-6, EUG-6 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P01730, P27487 | |
CD4, DPP4 | |
human peripheral blood T lymphocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1, IgG2a |