Learn More
CD4 Monoclonal Antibody (W3/25), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180603
Additional Details : Weight : 0.01000kg
Description
This antibody inhibits proliferation and IL-2 production in the MLR reaction. PLP fixation is recommended for optimal results. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD4 antibody, clone W3/25 recognizes the rat CD4 cell surface glycoprotein, a approximately 55 kDa molecule expressed by helper T cells and weakly by monocytes.
CD4 Monoclonal antibody specifically detects CD4 in Rat samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
CD4 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P05540 | |
CD4 | |
Rat Thymocyte Membrane Glycoproteins. | |
1 mg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
W3/25 | |
Unconjugated | |
CD4 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
Mouse | |
Protein A | |
RUO | |
24932 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |