Learn More
anti-CD3 zeta (FITC), Clone: H146-968, Invitrogen™
Specifically detect CD247 / CD3z (FITC) in Human, Mouse samples using Thermo Scientifc™ Pierce™ anti-CD247 / CD3z, FITC Conjugated Clone: H146-968, validated for FACS
Brand: Invitrogen MA110222
2023.43 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Specifically detect CD247 / CD3z (FITC) in Human, Mouse samples using Thermo Scientifc™ Pierce™ anti-CD247 / CD3z, FITC Conjugated Clone: H146-968, validated for FACS
The protein encoded by this gene is T-cell receptor zeta, which together with T-cell receptor alpha/beta and gamma/delta heterodimers, and with CD3-gamma, -delta and -epsilon, forms the T-cell receptor-CD3 complex. The zeta chain plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. Low expression of the antigen results in impaired immune response. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq, Jul 2008]
Host Species: Hamster
Cross Reactivity: Human, Mouse
Isotype: IgG
Clone: H146-968
Immunogen: Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta.
Formulation: 100μg of purified antibody in PBS buffer; pH 7.4.
Synonyms: RP11-104L21.1, CD3-ZETA, CD3H, CD3Q, CD3Z, T3Z, TCRZ, CD247 antigen, zeta subunit, CD3Z antigen, zeta polypeptide (TiT3 complex), T-cell antigen receptor complex, zeta subunit of CD3, T-cell receptor T3 zeta chain, T-cell surface glycoprotein CD3 zeta chain
Uniprot Accession: P20963, P29020
Entrez Gene ID: 919, 12503
MA1-10189 detects CD247 / CD3z in Human and Mouse samples. MA1-10189 has been successfully used in FACS procedures.MA1-10189 The immunogen is Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta.
Specifications
CD3 ζ | |
Monoclonal | |
FITC | |
Hamster | |
Unpurified | |
RUO | |
Store at 4°C. Store in the dark. DO NOT FREEZE. |
Flow Cytometry | |
H146-968 | |
CD247 | |
Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta. | |
100 μg | |
Primary | |
IgG |