Learn More
CD3e Monoclonal Antibody (SP7), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514524
Additional Details : Weight : 0.01000kg
Description
MA5-14524 targets CD3E in IHC (P), FACS, and Western blot applications and shows reactivity with human and mouse samples. The MA5-14524 immunogen is a synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein.
CD3e Monoclonal antibody specifically detects CD3e in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD3e | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 15mM sodium azide | |
P04234, P04235, P07766, P09693, P11942, P20963, P22646, P24161 | |
Cd247, Cd3d, CD3E, CD3g | |
A synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein | |
RUO | |
12500, 12501, 12502, 12503, 915, 916, 917, 919 | |
4° C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
SP7 | |
Unconjugated | |
CD3g | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Rabbit | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |