Learn More
CD2 Monoclonal Antibody (TS2/18), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA0200
Additional Details : Weight : 0.80000kg
Description
MA0200 targets CD2 in FACS, IA, IP, and WB applications and shows reactivity with Human samples. This antibody does not react with rat tissue in Western blot applications. The MA0200 immunogen is human LFA-2 (CD2). MA0200 detects CD2 which has a predicted molecular weight of approximately 37 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot.Specifications
CD2 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
P06729 | |
CD2 | |
Human LFA-2 (CD2) | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot | |
TS2/18 | |
Unconjugated | |
CD2 | |
Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
Mouse | |
Protein A | |
RUO | |
914 | |
-20°C | |
Liquid |